Please login to your online account to display your discounted pricing

1,1,2-Trimethyl-1H-benz[e]indole 96%, ACROS Organics™

$129.03 - $129.03

Chemical Identifiers

Chemical Name or Material 1, 1, 2-Trimethyl-1H-benz[e]indole
Name Note 96%
CAS 41532-84-7
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C15H15N
Formula Weight 209.29
MDL Number MFCD00082627
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC421900250

  SDS
thermo fisher scientific
421900250
Glass bottle 25g Each for $129.03
Description & Specifications

Specifications

Assay ≥95%
Physical Form Crystalline Powder
Formula Weight 209.29
Quantity 25g
Color Yellow to brown
TSCA TSCA
Melting Point 111° to 117°C
EINECS Number 255-429-8
Density 0.7g/mL
CAS Max % 100.0
CAS Min % 95.0
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Synonym 2,3,3-Trimethylnaphto[1,2-d]pyrrole
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Chemical Name or Material 1, 1, 2-Trimethyl-1H-benz[e]indole
Name Note 96%
Molecular Formula C15H15N
CAS 41532-84-7
MDL Number MFCD00082627
Assay Percent Range 96%
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.