Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,2-Trichloro-3,3,3-trifluoro-1-propene, 97%

Chemical Identifiers

Chemical Name or Material 1,1,2-Trichloro-3,3,3-trifluoro-1-propene
CAS 431-52-7
Molecular Formula F3CC(Cl)=CCl2
Formula Weight 199.39
MDL Number MFCD00013672
UN Number UN2810
Beilstein 1766481
Synonym FC-1213xa
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0926303

 
alfa aesar™
L0926303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0926306

 
alfa aesar™
L0926306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0926314

 
alfa aesar™
L0926314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.39
CAS 431-52-7
Chemical Name or Material 1,1,2-Trichloro-3,3,3-trifluoro-1-propene
Assay 97%
Molecular Formula F3CC(Cl)=CCl2
MDL Number MFCD00013672
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2810
Beilstein 1766481
Synonym FC-1213xa
Health Hazard 1 Danger
Boiling Point 88°C
Melting Point −114°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number UD2696000
Quantity 1g
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.