Please login to your online account to display your discounted pricing

(±)-1-(1,2-Diphenylethyl)piperidine maleate, Tocris


NMDA antagonist, acts ion channel site

Quantity 10mg

Manufacturer: r & d systems  RDS036010

 View more versions of this product

Catalog No. 036010

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 381.47
Assay Percent Range >98%
Molecular Formula C19H23N·C4H4O4
CAS 207461-99-2
Chemical Name or Material (±)-1-(1, 2-Diphenylethyl)piperidine maleate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (±)-1-(1, 2-Diphenylethyl)piperidine maleate
Assay Percent Range >98%
Molecular Formula C19H23N·C4H4O4
Formula Weight 381.47
Accessories

Store at Room Temperature