Please login to your online account to display your discounted pricing

(±)-1-(1,2-Diphenylethyl)piperidine maleate, Tocris

NMDA antagonist, acts ion channel site

$149.90 - $632.23

Chemical Identifiers

Chemical Name or Material (±)-1-(1, 2-Diphenylethyl)piperidine maleate
Assay Percent Range >98%
Molecular Formula C19H23N·C4H4O4
Formula Weight 381.47
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

036050

 
r & d systems
RDS036050
50mg Each for $632.23

036010

 
r & d systems
RDS036010
10mg Each for $149.90
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 381.47
Chemical Name or Material (±)-1-(1, 2-Diphenylethyl)piperidine maleate
Assay Percent Range >98%
Molecular Formula C19H23N·C4H4O4
CAS 207461-99-2
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature