Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,2,2-Tetramethoxycyclohexane, 98%

$79.72 - $310.39

Chemical Identifiers

Chemical Name or Material 1,1,2,2-Tetramethoxycyclohexane
CAS 163125-34-6
Molecular Formula C10H20O4
Formula Weight 204.27
MDL Number MFCD00798587
Synonym 1,2-Cyclohexanedione bis(dimethyl acetal)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2044406

  SDS
alfa aesar™
B2044406
5g Each for $79.72

AAB2044414

  SDS
alfa aesar™
B2044414
25g Each for $310.39
Description & Specifications

Specifications

Formula Weight 204.27
Molecular Formula C10H20O4
CAS 163125-34-6
Quantity 5g
Chemical Name or Material 1,1,2,2-Tetramethoxycyclohexane
Assay 98%
Synonym 1,2-Cyclohexanedione bis(dimethyl acetal)
MDL Number MFCD00798587
Boiling Point 54°C (0.4mmHg)