Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,2,2-Tetrafluoroethyl methyl ether, 99%

Chemical Identifiers

Chemical Name or Material 1,1,2,2-Tetrafluoroethyl methyl ether
CAS 425-88-7
Molecular Formula CH3OCF2CHF2
Formula Weight 132.06
MDL Number MFCD00042106
UN Number UN3271
Beilstein 1737755
Synonym Methyl 1,1,2,2-tetrafluoroethyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1788906

  SDS
alfa aesar™
L1788906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1788914

  SDS
alfa aesar™
L1788914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 132.06
Assay 99%
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
CAS 425-88-7
MDL Number MFCD00042106
Chemical Name or Material 1,1,2,2-Tetrafluoroethyl methyl ether
UN Number UN3271
Melting Point −107°C
Boiling Point 36° to 37°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
Health Hazard 1 Danger
Synonym Methyl 1,1,2,2-tetrafluoroethyl ether
Quantity 5g
Beilstein 1737755
Molecular Formula CH3OCF2CHF2
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.