missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2,2-Tetrafluoroethyl 2,2,3,3-Tetrafluoropropyl Ether, TCI America™

Manufacturer:  TCI America T306925G

 View more versions of this product

Catalog No. T306925G



Specifications

Specifications

1,1,2,2-Tetrafluoroethyl 2,2,3,3-Tetrafluoropropyl Ether
Colorless
16627-68-2
MFCD00155961
HCBRSIIGBBDDCD-UHFFFAOYSA-N
1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane
2776662
Clear Liquid at 20°C
92°C
25g
C5H4F8O
3271
C(C(C(F)F)(F)F)OC(C(F)F)(F)F
232.073
232.07
≥98.0% (GC)

Chemical Identifiers

16627-68-2
232.073
HCBRSIIGBBDDCD-UHFFFAOYSA-N
1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane
C5H4F8O
MFCD00155961
2776662
C(C(C(F)F)(F)F)OC(C(F)F)(F)F
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4074

TSCA : No

Documents