missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2,2-Tetrafluoroethyl 2,2,2-Trifluoroethyl Ether, TCI America™

Manufacturer:  TCI America T305725G

 View more versions of this product

Catalog No. T305725G



Specifications

Specifications

1,1,2,2-Tetrafluoroethyl 2,2,2-Trifluoroethyl Ether
Yellow
406-78-0
MFCD00236118
CWIFAKBLLXGZIC-UHFFFAOYSA-N
1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane
164596
Clear Liquid at 20°C
56°C
25g
C4H3F7O
3271
C(C(F)(F)F)OC(C(F)F)(F)F
200.056
200.06
≥99.0% (GC)

Chemical Identifiers

406-78-0
200.056
CWIFAKBLLXGZIC-UHFFFAOYSA-N
1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane
C4H3F7O
MFCD00236118
164596
C(C(F)(F)F)OC(C(F)F)(F)F
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3983

TSCA : Yes

Documents