Please login to your online account to display your discounted pricing

1,1,2,2-Tetrachloroethane-d2, for NMR, 99.5+ atom% D, ACROS Organics™

$38.89 - $38.89

Chemical Identifiers

Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Name Note 99.5+ atom% D, for NMR
CAS 33685-54-0
Purity Grade For NMR
Molecular Formula C2Cl4D2
Linear Formula (Cl2CD)2
Formula Weight 169.86
MDL Number MFCD00037672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC321310010

  SDS
thermo fisher scientific
321310010
Glass bottle 1g Each for $38.89
Description & Specifications

Specifications

Formula Weight 169.86
Physical Form Liquid
Quantity 1g
Color Clear colorless
Purity Grade For NMR
Density 1.6100g/mL
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Packaging Glass bottle
Linear Formula (Cl2CD)2
CAS 33685-54-0
Molecular Formula C2Cl4D2
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00037672
Deuterium (D) 99.50% min.
Name Note 99.5+ atom% D, for NMR
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.