Please login to your online account to display your discounted pricing

1,1,2,2 - Tetrachloroethane - D2, deuteration deg. min. 99.5% for NMR, MagniSolv™, EMD Millipore™


Quantity 10 x 0.75mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  1034950009

 View more versions of this product

Catalog No. M1034950009

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 1,1,2,2 - Tetrachloroethane - D2
CAS 33685-54-0
Odor Sweet
Packaging Glass ampoule
Vapor Pressure 6.6 hPa at 20°C (68°F)
Viscosity 1.84mPa&middnot;s
Physical Form Liquid
Quantity 10 x 0.75mL
Assay Percent Range 99.5% min.
Color Colorless
Molecular Formula C2D2Cl4
Formula Weight 169.86
Synonym 1,2 - Dideutero - 1,1,2,2 - tetrachloroethane

Specifications

Chemical Name or Material 1,1,2,2 - Tetrachloroethane - D2
CAS 33685-54-0
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C2D2Cl4
Formula Weight 169.86
Synonym 1,2 - Dideutero - 1,1,2,2 - tetrachloroethane