missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2,2 - Tetrachloroethane - D2, deuteration deg. min. 99.5% for NMR, MagniSolv™, EMD Millipore™

$267.95 - $591.16

Chemical Identifiers

CAS 33685-54-0
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C2D2Cl4
Formula Weight 169.86
Synonym 1,2 - Dideutero - 1,1,2,2 - tetrachloroethane
Chemical Name or Material 1,1,2,2 - Tetrachloroethane - D2
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M1034950009 Emd Millipore Corporation
1034950009
Glass ampoule 10 x 0.75mL Each for $267.95
M1034950025 Emd Millipore Corporation
1034950025
Glass Bottle 25mL Each for $591.16
Specifications Description & Specifications

Chemical Identifiers

33685-54-0
99.5% min.
169.86
1,1,2,2 - Tetrachloroethane - D2
Liquid
C2D2Cl4
1,2 - Dideutero - 1,1,2,2 - tetrachloroethane

Specifications

33685-54-0
Glass ampoule
1.84mPa&middnot;s
10 x 0.75mL
Colorless
169.86
1,1,2,2 - Tetrachloroethane - D2
Sweet
6.6 hPa at 20°C (68°F)
Liquid
99.5% min.
C2D2Cl4
1,2 - Dideutero - 1,1,2,2 - tetrachloroethane