Please login to your online account to display your discounted pricing

1,1,2,2-Tetrachloroethane-d2 99 atom% D, ACROS Organics™

$108.70 - $295.16

Chemical Identifiers

Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Name Note 99 atom % D
CAS 33685-54-0
Purity Grade For NMR
Molecular Formula C2Cl4D2
Linear Formula (Cl2CD)2
Formula Weight 169.86
MDL Number MFCD00037672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC133580200

  SDS
thermo fisher scientific
133580200
Glass bottle 20mL Each for $295.16

AC133580050

  SDS
thermo fisher scientific
133580050
Glass bottle 5mL Each for $108.70
Description & Specifications

Specifications

Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Name Note 99 atom % D
CAS 33685-54-0
Deuterium (D) 98.5% min.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
EINECS Number 251-634-1
TSCA TSCA
Density 1.6100g/mL
Purity Grade For NMR
Physical Form Liquid
Quantity 20mL
Color Clear colorless
Molecular Formula C2Cl4D2
Linear Formula (Cl2CD)2
Formula Weight 169.86
MDL Number MFCD00037672
Beilstein 01, III, 164

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.