Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,2,2-Tetrachloroethane-d2, 99.6% (Isotopic)

$38.34 - $38.34

Chemical Identifiers

Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
CAS 33685-54-0
Name Note Isotopic
Linear Formula Cl2CDCDCl2
Formula Weight 169.86
MDL Number MFCD00037672
UN Number UN1702
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4234704

 
alfa aesar™
4234704*
2g Each for $38.34

AA4234718

 
alfa aesar™
4234718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.86
Health Hazard 3 GHS P Statement
P260-P304+P340-P320-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2
Density 1.620g/mL
Name Note Isotopic
Boiling Point 147°C
Melting Point −44°C
Physical Form Liquid
Assay 99.6%
Quantity 2g
Color Colorless
Linear Formula Cl2CDCDCl2
Refractive Index 1.4930
MDL Number MFCD00037672
CAS 33685-54-0
UN Number UN1702
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H310-H330
Fatal in contact with skin.
Fatal if inhaled.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,1,2,2-TETRACHLOROETHANE
Safety and Handling

Danger

GHS P Statement
P260-P304+P340-P320-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H310-H330
Fatal in contact with skin.
Fatal if inhaled.