Learn More
1,1,2,2-Tetrachloroethane, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0448936
Description
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.
Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
Specifications
1,1,2,2-Tetrachloroethane | |
-43°C | |
142°C to 146°C | |
1.4935 | |
MFCD00000848 | |
UN1702 | |
14,9189 | |
Miscible with water,methanol,ethanol,chloroform and acetone. | |
C(C(Cl)Cl)(Cl)Cl | |
167.838 | |
CHEBI:36026 | |
≥98% |
79-34-5 | |
1.596 | |
C2H2Cl4 | |
Cl2CHCHCl2 | |
500 g | |
969206 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
1,1,2,2-tetrachloroethane | |
6591 | |
167.85 |
Chemical Identifiers
79-34-5 | |
167.838 | |
QPFMBZIOSGYJDE-UHFFFAOYSA-N | |
6591 | |
1,1,2,2-tetrachloroethane |
C2H2Cl4 | |
MFCD00000848 | |
s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan | |
CHEBI:36026 | |
C(C(Cl)Cl)(Cl)Cl |
Safety and Handling
GHS H Statement
H310-H330-H341-H351
Fatal in contact with skin.
Fatal if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
P201-P202-P260-P262-P264b-P270-P271-P281-P284-P302+P350-P304+P340-P308+P313-P310-P361-P363-P501c
H310+H330-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,1,2,2-TETRACHLOROETHANE
EINECSNumber : 201-197-8
RTECSNumber : KI8575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only