missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,2,2-Tetrachloro-1,2-dimethyldisilane, TCI America™

Manufacturer:  TCI America T159025G

 View more versions of this product

Catalog No. T159025G



Specifications

Specifications

1,1,2,2-Tetrachloro-1,2-dimethyldisilane
25g
C2H6Cl4Si2
2988
C[Si]([Si](C)(Cl)Cl)(Cl)Cl
228.04
228.04
≥98.0% (T)
Colorless
4518-98-3
MFCD00192469
JTBAMRDUGCDKMS-UHFFFAOYSA-N
dichloro-[dichloro(methyl)silyl]-methylsilane
78276
Clear Liquid at 20°C

Chemical Identifiers

4518-98-3
228.04
JTBAMRDUGCDKMS-UHFFFAOYSA-N
dichloro-[dichloro(methyl)silyl]-methylsilane
C2H6Cl4Si2
MFCD00192469
78276
C[Si]([Si](C)(Cl)Cl)(Cl)Cl
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2040

TSCA : Yes

Recommended Storage : Refrigerator

Documents