Learn More
1,1,2,2-Tetrabromoethane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180870010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,1,2,2-Tetrabromoethane | |
97.5 | |
1.0°C | |
244.0°C | |
97.5% min. (GC) | |
C2H2Br4 | |
Br2CHCHBr2 | |
1 kg | |
2.96 | |
acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o | |
QXSZNDIIPUOQMB-UHFFFAOYSA-N | |
1,1,2,2-tetrabromoethane | |
6588 | |
98% |
79-27-6 | |
100.0 | |
2.9600g/mL | |
Authentic | |
Glass bottle | |
1.6355 to 1.6380 | |
MFCD00000133 | |
01, 94 | |
15, 9330 | |
Solubility in water: 0.63g/L (20°C). Other solubilities: soluble in acetone, benzene, miscible with ethanol, ether, chloroform, aniline, and glacial acetic acid | |
BrC(Br)C(Br)Br | |
345.65 | |
345.64 | |
Liquid |
Chemical Identifiers
79-27-6 | |
345.65 | |
QXSZNDIIPUOQMB-UHFFFAOYSA-N | |
6588 | |
BrC(Br)C(Br)Br |
C2H2Br4 | |
MFCD00000133 | |
acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o | |
1,1,2,2-tetrabromoethane |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-191-5
RUO – Research Use Only