Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1-Tris(diphenylphosphinomethyl)ethane, ≥97%

$238.07 - $238.07

Chemical Identifiers

Chemical Name or Material 1,1,1-Tris(diphenylphosphinomethyl)ethane
CAS 22031-12-5
Formula Weight 624.67
MDL Number MFCD00014084
Synonym Phenyl triphos; 1,3-Bis(diphenylphosphino)-2-(diphenylphosphino)methyl-2-methylpropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3786303

  SDS
alfa aesar™
H3786303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3786306

  SDS
alfa aesar™
H3786306
5g Each for $238.07
Description & Specifications

Specifications

Formula Weight 624.67
Melting Point 99° to 102°C
Assay ≥97%
Synonym Phenyl triphos; 1,3-Bis(diphenylphosphino)-2-(diphenylphosphino)methyl-2-methylpropane
Sensitivity Air Sensitive
Chemical Name or Material 1,1,1-Tris(diphenylphosphinomethyl)ethane
Physical Form Powder
CAS 22031-12-5
Color White
MDL Number MFCD00014084
Quantity 1g

Applications
1,1,1-Tris(diphenylphosphinomethyl)ethane is used in chemicals and pharmaceutical research.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.