Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1-Tris(chloromethyl)ethane, 98%

$161.23 - $497.23

Chemical Identifiers

Chemical Name or Material 1,1,1-Tris(chloromethyl)ethane
CAS 1067-09-0
Molecular Formula CH3C(CH2Cl)3
Formula Weight 175.49
MDL Number MFCD00013686
Synonym 1,3-Dichloro-2-chloromethyl-2-methylpropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1751414

  SDS
alfa aesar™
A1751414
25g Each for $161.23

AAA1751422

  SDS
alfa aesar™
A1751422
100g Each for $497.23
Description & Specifications

Specifications

Formula Weight 175.49
MDL Number MFCD00013686
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,1,1-Tris(chloromethyl)ethane
CAS 1067-09-0
Flash Point 86°C (187°F)
Boiling Point 85° to 87°C (14mmHg)
Quantity 25g
Melting Point 18°C
Molecular Formula CH3C(CH2Cl)3
Health Hazard 1 Warning
Assay 98%
Synonym 1,3-Dichloro-2-chloromethyl-2-methylpropane
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.