Learn More
1,1,1-Trifluoroacetone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139740250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 1, 1-Trifluoroacetone | |
-78.0°C | |
1.2500g/mL | |
−30°C | |
98% min. (GC) | |
C3H3F3O | |
CF3COCH3 | |
01,II,717 | |
1.25 | |
Solubility in water: miscible | |
CC(=O)C(F)(F)F | |
112.05 | |
3.28 mPa.s (15°C) | |
98+% |
421-50-1 | |
Colorless | |
22.0°C | |
Authentic | |
Glass bottle | |
1.3000 (20°C, 589nm) max. | |
25 g | |
13,323; 15,212 | |
1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone | |
FHUDAMLDXFJHJE-UHFFFAOYSA-N | |
1,1,1-trifluoropropan-2-one | |
9871 | |
112.05 | |
Liquid |
Chemical Identifiers
421-50-1 | |
112.05 | |
1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone | |
1,1,1-trifluoropropan-2-one |
C3H3F3O | |
FHUDAMLDXFJHJE-UHFFFAOYSA-N | |
9871 | |
CC(=O)C(F)(F)F |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 207-005-9
RUO – Research Use Only