Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1-Trifluoroacetone, 95%

Chemical Identifiers

Chemical Name or Material 1,1,1-Trifluoroacetone
CAS 421-50-1
Molecular Formula CF3COCH3
Formula Weight 112.05
MDL Number MFCD00000423
UN Number UN1993
Beilstein 1748614
Synonym 1,1,1-Trifluoro-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1355622

  SDS
alfa aesar™
A1355622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.05
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
Odor Chloroform-like
TSCA TSCA
CAS 421-50-1
Flash Point −31°C (−24°F)
Melting Point <−78°C
Molecular Formula CF3COCH3
MDL Number MFCD00000423
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Synonym 1,1,1-Trifluoro-2-propanone
UN Number UN1993
Boiling Point 21° to 24°C
Chemical Name or Material 1,1,1-Trifluoroacetone
Beilstein 1748614
Assay 95%
Quantity 100g

Applications
1,1,1-Trifluoroacetone is used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines. The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a Strecker reaction followed by either nitrile hydrolysis or reduction. And also used as a useful molecule in synthesis reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C