missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,1-Trifluoro-4-iodobutane, TCI America™

Manufacturer:  TCI America T252925G

 View more versions of this product

Catalog No. T252925G



Specifications

Specifications

1,1,1-Trifluoro-4-iodobutane
Undesignated
461-17-6
MFCD00041543
LNDGACQEAYKNOI-UHFFFAOYSA-N
1,1,1-trifluoro-4-iodobutane
2775189
Clear Liquid at 20°C
128°C
25g
C4H6F3I
1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
C(CC(F)(F)F)CI
237.992
237.99
≥98.0% (GC)

Chemical Identifiers

461-17-6
237.992
LNDGACQEAYKNOI-UHFFFAOYSA-N
2775189
C(CC(F)(F)F)CI
C4H6F3I
MFCD00041543
1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane
1,1,1-trifluoro-4-iodobutane
Safety and Handling

Safety and Handling

TSCA : No

Documents