Learn More
1,1,1-Trifluoro-2-propanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1115014
Description
1,1,1-Trifluoro-2-propanol is used as a chemical building block.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,1-Trifluoro-2-propanol is used as a chemical building block.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications
1,1,1-Trifluoro-2-propanol | |
1.259 | |
25°C (77°F) | |
1.3175 | |
25 g | |
1734738 | |
Fully miscible in water. | |
CC(C(F)(F)F)O | |
114.067 | |
114.07 |
374-01-6 | |
75°C to 76°C | |
C3H5F3O | |
MFCD00014403 | |
UN1987 | |
1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro | |
GILIYJDBJZWGBG-UHFFFAOYSA-N | |
1,1,1-trifluoropropan-2-ol | |
9774 | |
97% |
Chemical Identifiers
374-01-6 | |
114.067 | |
GILIYJDBJZWGBG-UHFFFAOYSA-N | |
9774 | |
CC(C(F)(F)F)O |
C3H5F3O | |
MFCD00014403 | |
1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro | |
1,1,1-trifluoropropan-2-ol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 206-773-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only