Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1-Trifluoro-2-phenyl-3-butyn-2-ol, 96%

$191.36 - $792.55

Chemical Identifiers

Chemical Name or Material 1,1,1-Trifluoro-2-phenyl-3-butyn-2-ol
CAS 99727-20-5
Molecular Formula C10H7F3O
Formula Weight 200.16
MDL Number MFCD00792447
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5109903

  SDS
alfa aesar™
H5109903
1g Each for $191.36

AAH5109906

  SDS
alfa aesar™
H5109906
5g Each for $792.55
Description & Specifications

Specifications

Formula Weight 200.16
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,1,1-Trifluoro-2-phenyl-3-butyn-2-ol
Assay 96%
Molecular Formula C10H7F3O
MDL Number MFCD00792447
CAS 99727-20-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.