missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1-Trifluoro-2,4-pentanedione, 98%

Manufacturer:  Alfa Aesar™ A1468206

 View more versions of this product

Catalog No. AAA1468206



Description

Description

Applications
1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

Solubility
Soluble in water (slightly) and many organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1,1,1-Trifluoro-2,4-pentanedione
105°C to 107°C
367-57-7
MFCD00000427
1705177
SHXHPUAKLCCLDV-UHFFFAOYSA-N
1,1,1-trifluoropentane-2,4-dione
73943
98%
26°C (79°F)
5g
C5H5F3O2
UN1224
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
CC(=O)CC(=O)C(F)(F)F
154.088
154.09

Chemical Identifiers

367-57-7
154.088
SHXHPUAKLCCLDV-UHFFFAOYSA-N
73943
CC(=O)CC(=O)C(F)(F)F
C5H5F3O2
MFCD00000427
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
1,1,1-trifluoropentane-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA

Documents