Learn More
1,1,1-Trifluoro-2,4-hexanedione, 70%, tech., ACROS Organics™
Chemical Identifiers
CAS | 400-54-4 |
---|---|
Molecular Formula | C6H7F3O2 |
Molecular Weight (g/mol) | 168.12 |
MDL Number | MFCD00051306 |
InChI Key | SBRYTXXHTKCMFT-UHFFFAOYSA-N |
Synonym | 1,1,1,-trifluoro-2,4-hexanedione, 1,1,1-trifluoro-2,4-hexane-dione, 1,1,1-trifluoro-2,4-hexanedione, 1,1,1-trifluoro-hexane-2,4-dione, 1,1,1-trifluorohexan-2,4-dione, 1-trifluoroacetyl butan-2-one, 2,4-dioxo-1,1,1-trifluorohexane, 2,4-hexanedione, 1,1,1-trifluoro, 2,4-hexanedione,1,1,1-trifluoro |
PubChem CID | 67864 |
IUPAC Name | 1,1,1-trifluorohexane-2,4-dione |
SMILES | CCC(=O)CC(=O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC421750250
|
Thermo Fisher Scientific
421750250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC421751000
|
Thermo Fisher Scientific
AC421751000 |
100g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
400-54-4 | |
168.12 | |
SBRYTXXHTKCMFT-UHFFFAOYSA-N | |
67864 | |
CCC(=O)CC(=O)C(F)(F)F |
C6H7F3O2 | |
MFCD00051306 | |
1,1,1,-trifluoro-2,4-hexanedione, 1,1,1-trifluoro-2,4-hexane-dione, 1,1,1-trifluoro-2,4-hexanedione, 1,1,1-trifluoro-hexane-2,4-dione, 1,1,1-trifluorohexan-2,4-dione, 1-trifluoroacetyl butan-2-one, 2,4-dioxo-1,1,1-trifluorohexane, 2,4-hexanedione, 1,1,1-trifluoro, 2,4-hexanedione,1,1,1-trifluoro | |
1,1,1-trifluorohexane-2,4-dione |
Specifications
28g/mL | |
Authentic | |
1.3931 to 1.3951 | |
Yellow | |
400-54-4 | |
100.0 | |
C6H7F3O2 | |
MFCD00051306 | |
SBRYTXXHTKCMFT-UHFFFAOYSA-N | |
1,1,1-trifluorohexane-2,4-dione | |
67864 | |
Liquid | |
Technical |
28°C | |
Glass bottle | |
125.0°C | |
25g | |
65.0 | |
65% min. (GC) | |
CF3COCH2COCH2CH3 | |
1,1,1,-trifluoro-2,4-hexanedione, 1,1,1-trifluoro-2,4-hexane-dione, 1,1,1-trifluoro-2,4-hexanedione, 1,1,1-trifluoro-hexane-2,4-dione, 1,1,1-trifluorohexan-2,4-dione, 1-trifluoroacetyl butan-2-one, 2,4-dioxo-1,1,1-trifluorohexane, 2,4-hexanedione, 1,1,1-trifluoro, 2,4-hexanedione,1,1,1-trifluoro | |
CCC(=O)CC(=O)C(F)(F)F | |
168.12 | |
168.12 | |
70% | |
1,1,1-Trifluoro-2,4-hexanedione |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 206-924-2