Please login to your online account to display your discounted pricing

1,1,1-Trifluoro-2,4-hexanedione, 70%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  421750250

 View more versions of this product

Catalog No. AC421750250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥65%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 168.12
Quantity 25g
Color Clear yellow
Density 28g/mL
CAS Min % 65.0
CAS Max % 100.0
Linear Formula CF3COCH2COCH2CH3
Chemical Name or Material 1,1,1-Trifluoro-2,4-hexanedione
Infrared Spectrum Authentic
Assay Percent Range 70%
Molecular Formula C6H7F3O2
MDL Number MFCD00051306
CAS 400-54-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material 1,1,1-Trifluoro-2,4-hexanedione
CAS 400-54-4
CAS Min % 65.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥65%
Assay Percent Range 70%
Molecular Formula C6H7F3O2
Linear Formula CF3COCH2COCH2CH3
Formula Weight 168.12
MDL Number MFCD00051306
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Flammable liquid and vapor.

SDS & Safety Documents