missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150305000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 1, 1-Trichloro-2-methyl-2-propanol hemihydrate | |
6001-64-5 | |
100.0 | |
White | |
100°C | |
97.5 to 102.5% (ex Cl) (Argentometry) | |
0·5 H2O | |
MFCD02179352 | |
c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate | |
HBARVHNVVKZUGS-UHFFFAOYSA-N | |
molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate | |
102594540 | |
98% |
98% | |
97.5 | |
75°C to 79°C | |
173°C to 175°C | |
Authentic | |
Plastic bottle | |
0·5H2O | |
500 g | |
Solubility in water: 7.7g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,glycerol | |
[HH].CC(C)(C(Cl)(Cl)Cl)O.O | |
186.47 | |
186.47 | |
Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
6001-64-5 | |
186.47 | |
HBARVHNVVKZUGS-UHFFFAOYSA-N | |
102594540 | |
[HH].CC(C)(C(Cl)(Cl)Cl)O.O |
0·5 H2O | |
MFCD02179352 | |
c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate | |
molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only