missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione, 97%

$213.96 - $213.96

Chemical Identifiers

CAS 20583-66-8
Molecular Formula C7H2F10O2
Formula Weight 308.08
MDL Number MFCD00040961
UN Number UN3265
Beilstein 1889488
Chemical Name or Material 1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0972403 SDS Alfa Aesar™
L0972403
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL0972406 SDS Alfa Aesar™
L0972406
5g Each for $213.96
AAL0972414 SDS Alfa Aesar™
L0972414
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

20583-66-8
308.08
UN3265
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
C7H2F10O2
MFCD00040961
1889488

Specifications

20583-66-8
98° to 100°C
MFCD00040961
1889488
1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
C7H2F10O2
308.08
UN3265
1g
97%

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.