missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione, 97%

Manufacturer:  Alfa Aesar™ L0972403

 View more versions of this product

Catalog No. AAL0972403



Specifications

Specifications

1,1,1,5,5,6,6,7,7,7-Decafluoro-2,4-heptanedione
1g
C7H2F10O2
UN3265
1,1,1,5,5,6,6,7,7,7-decafluoro-2,4-heptanedione, 3h,3h-perfluoroheptane-2,4-dione, 1,1,1,5,5,6,6,7,7,7-decafluoroheptane-2,4-dione, acmc-1cb0q, 3h,3h-perfluoro-2,4-heptanedione, 1,1,1,2,2,3,3,7,7,7-decafluoroheptane-4,6-dione, 1,1,1,5,5,6,6,7,7,7-decafluoro-2,4-heptadione, 2,4-heptanedione,1,1,1,5,5,6,6,7,7,7-decafluoro, 1,1,1,5,5,6,6,7,7,7-decakis fluoranyl heptane-2,4-dione
C(C(=O)C(C(C(F)(F)F)(F)F)(F)F)C(=O)C(F)(F)F
308.075
308.08
98°C to 100°C
20583-66-8
MFCD00040961
1889488
SUORUQZBFOQDGX-UHFFFAOYSA-N
1,1,1,5,5,6,6,7,7,7-decafluoroheptane-2,4-dione
425461
97%

Chemical Identifiers

20583-66-8
308.075
SUORUQZBFOQDGX-UHFFFAOYSA-N
425461
C(C(=O)C(C(C(F)(F)F)(F)F)(F)F)C(=O)C(F)(F)F
C7H2F10O2
MFCD00040961
1,1,1,5,5,6,6,7,7,7-decafluoro-2,4-heptanedione, 3h,3h-perfluoroheptane-2,4-dione, 1,1,1,5,5,6,6,7,7,7-decafluoroheptane-2,4-dione, acmc-1cb0q, 3h,3h-perfluoro-2,4-heptanedione, 1,1,1,2,2,3,3,7,7,7-decafluoroheptane-4,6-dione, 1,1,1,5,5,6,6,7,7,7-decafluoro-2,4-heptadione, 2,4-heptanedione,1,1,1,5,5,6,6,7,7,7-decafluoro, 1,1,1,5,5,6,6,7,7,7-decakis fluoranyl heptane-2,4-dione
1,1,1,5,5,6,6,7,7,7-decafluoroheptane-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents