Please login to your online account to display your discounted pricing

1,1,1,5,5,5-Hexafluoroacetylacetone, 99%, ACROS Organics™

$59.49 - $205.27

Chemical Identifiers

Chemical Name or Material 1,1,1,5,5,5-Hexafluoroacetylacetone
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H2F6O2
Linear Formula CF3COCH2COCF3
Formula Weight 208.06
Synonym 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC218530050

  SDS
thermo fisher scientific
218530050
Glass bottle 5g Each for $59.49

AC218531000

  SDS
thermo fisher scientific
AC218531000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC218530250

  SDS
thermo fisher scientific
AC218530250
Glass bottle 25g Each for $205.27
Description & Specifications

Specifications

Assay ≥98.5% (GC)
Formula Weight 208.06
Physical Form Liquid
Quantity 5g
Color Clear colorless to slightly yellow
Density 1.47g/mL
CAS 1522-22-1
Assay Percent Range 99%
Synonym 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione
Linear Formula CF3COCH2COCF3
Molecular Formula C5H2F6O2
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3320 to 1.3340 (20°C, 589nm)
Chemical Name or Material 1,1,1,5,5,5-Hexafluoroacetylacetone