missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%

$66.74 - $599.26

Chemical Identifiers

CAS 1522-22-1
Molecular Formula C5H2F6O2
Molecular Weight (g/mol) 208.059
MDL Number MFCD00000426
InChI Key QAMFBRUWYYMMGJ-UHFFFAOYSA-N
Synonym 1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 1,3-bis trifluoromethyl propane-1,3-dione, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hexafluoroacetylacetone, hfac
PubChem CID 73706
IUPAC Name 1,1,1,5,5,5-hexafluoropentane-2,4-dione
SMILES C(C(=O)C(F)(F)F)C(=O)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0169306 View Documents Alfa Aesar™
L0169306
5g Each for $66.74
 
AAL0169314 View Documents Alfa Aesar™
L0169314
25g Each for $198.08
 
AAL0169322 View Documents Alfa Aesar™
L0169322
100g Each for $599.26
 
Description

Description

Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
Specifications

Chemical Identifiers

1522-22-1
208.059
QAMFBRUWYYMMGJ-UHFFFAOYSA-N
73706
C(C(=O)C(F)(F)F)C(=O)C(F)(F)F
C5H2F6O2
MFCD00000426
1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 1,3-bis trifluoromethyl propane-1,3-dione, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hexafluoroacetylacetone, hfac
1,1,1,5,5,5-hexafluoropentane-2,4-dione

Specifications

1.486
1.334
69°C to 71°C
1522-22-1
MFCD00000426
1790138
Not miscible in water.
C(C(=O)C(F)(F)F)C(=O)C(F)(F)F
208.059
208.06
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione
32°C (89°F)
Hygroscopic
5g
C5H2F6O2
UN2920
1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 1,3-bis trifluoromethyl propane-1,3-dione, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hexafluoroacetylacetone, hfac
QAMFBRUWYYMMGJ-UHFFFAOYSA-N
1,1,1,5,5,5-hexafluoropentane-2,4-dione
73706
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 216-191-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen