missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%

Manufacturer:  Alfa Aesar™ L0169314

 View more versions of this product

Catalog No. AAL0169314



Description

Description

Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
Specifications

Specifications

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione
32°C (89°F)
Hygroscopic
25g
C5H2F6O2
UN2920
1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 1,3-bis trifluoromethyl propane-1,3-dione, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hexafluoroacetylacetone, hfac
QAMFBRUWYYMMGJ-UHFFFAOYSA-N
1,1,1,5,5,5-hexafluoropentane-2,4-dione
73706
≥98%
1.486
1.334
69°C to 71°C
1522-22-1
MFCD00000426
1790138
Not miscible in water.
C(C(=O)C(F)(F)F)C(=O)C(F)(F)F
208.059
208.06

Chemical Identifiers

1522-22-1
208.059
QAMFBRUWYYMMGJ-UHFFFAOYSA-N
73706
C(C(=O)C(F)(F)F)C(=O)C(F)(F)F
C5H2F6O2
MFCD00000426
1,1,1,5,5,5-hexafluoro-2,4-pentadione, 1,1,1,5,5,5-hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-hexafluoroacetylacetone, 1,1,1-trifluoro-3-trifluoroacetyl acetone, 1,3-bis trifluoromethyl propane-1,3-dione, 2,4-dioxo-1,1,1,5,5,5-hexafluoropentane, 2,4-pentanedione, 1,1,1,5,5,5-hexafluoro, hexafluoro-2,4-pentanedione, hexafluoroacetylacetone, hfac
1,1,1,5,5,5-hexafluoropentane-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 216-191-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents
Product Certifications