missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,1,3,3-Pentamethyldisiloxane, TCI America™

Manufacturer:  TCI America P153525ML

 View more versions of this product

Catalog No. P153525ML



Specifications

Specifications

1,1,1,3,3-Pentamethyldisiloxane
Colorless
1438-82-0
MFCD00053658
pentamethyldisiloxane, disiloxane, pentamethyl, 1,1,1,3,3-pentamethyldisiloxane, dimethyl trimethylsilyloxy silicon, disiloxane, 1,1,1,3,3-pentamethyl, dimethyl trimethylsilyl oxy silyl, dimethylsilyl oxy trimethylsilane
C[Si](C)O[Si](C)(C)C
147.344
148.35
≥95.0% (GC)
86°C
25mL
C5H15OSi2
1993
XUKFPAQLGOOCNJ-UHFFFAOYSA-N
dimethyl(trimethylsilyloxy)silicon
6327260
Clear Liquid at 20°C

Chemical Identifiers

1438-82-0
147.344
XUKFPAQLGOOCNJ-UHFFFAOYSA-N
6327260
C[Si](C)O[Si](C)(C)C
C5H15OSi2
MFCD00053658
pentamethyldisiloxane, disiloxane, pentamethyl, 1,1,1,3,3-pentamethyldisiloxane, dimethyl trimethylsilyloxy silicon, disiloxane, 1,1,1,3,3-pentamethyl, dimethyl trimethylsilyl oxy silyl, dimethylsilyl oxy trimethylsilane
dimethyl(trimethylsilyloxy)silicon
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0477

TSCA : Yes

Documents