missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate, 97%

Manufacturer:  Alfa Aesar™ L1086414

 View more versions of this product

Catalog No. AAL1086414



Specifications

Specifications

1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate
25g
C10H8F6O3S
4530866
QGSVBFAODCJVIZ-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzenesulfonate
98916
97%
39°C to 40°C
67674-48-0
MFCD00039262
1,1,1,3,3,3-hexafluoroisopropyl p-toluenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 2-4-methylbenzenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl 4-toluenesulphonate, 2,2,2-trifluoro-1-trifluoromethyl ethyl toluene-p-sulphonate, hexafluoroisopropyl 4-toluenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl tosylate, 1,1,1,3,3,3-hexafluoroisopropyl-4-toluensesulfonate, 2-propanol,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, p-toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F
322.221
322.23

Chemical Identifiers

67674-48-0
322.221
QGSVBFAODCJVIZ-UHFFFAOYSA-N
98916
CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F
C10H8F6O3S
MFCD00039262
1,1,1,3,3,3-hexafluoroisopropyl p-toluenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 2-4-methylbenzenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl 4-toluenesulphonate, 2,2,2-trifluoro-1-trifluoromethyl ethyl toluene-p-sulphonate, hexafluoroisopropyl 4-toluenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl tosylate, 1,1,1,3,3,3-hexafluoroisopropyl-4-toluensesulfonate, 2-propanol,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, p-toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents