Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate, 97%

$50.78 - $153.69

Chemical Identifiers

Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate
CAS 67674-48-0
Molecular Formula C10H8F6O3S
Formula Weight 322.23
MDL Number MFCD00039262
Beilstein 4530866
Synonym p-Toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1086406

  SDS
alfa aesar™
L1086406
5g Each for $50.78

AAL1086414

  SDS
alfa aesar™
L1086414
25g Each for $153.69
Description & Specifications

Specifications

Formula Weight 322.23
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl p-toluenesulfonate
Assay 97%
MDL Number MFCD00039262
Synonym p-Toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
CAS 67674-48-0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H8F6O3S
TSCA TSCA
Quantity 5g
Beilstein 4530866
Melting Point 39° to 40°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.