missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,1,1,3,3,3-Hexafluoroisopropyl Methacrylate (stabilized with MEHQ), TCI America™

$100.83 - $200.14

Chemical Identifiers

CAS 3063-94-3
Molecular Formula C7H6F6O2
Molecular Weight (g/mol) 236.113
MDL Number MFCD00040105
InChI Key FMQPBWHSNCRVQJ-UHFFFAOYSA-N
Synonym hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
PubChem CID 76469
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M098810G View Documents TCI America
M098810G
10g Each for $100.83
 
M098825G View Documents TCI America
M098825G
25g Each for $200.14
 
Specifications

Chemical Identifiers

3063-94-3
236.113
FMQPBWHSNCRVQJ-UHFFFAOYSA-N
76469
CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
C7H6F6O2
MFCD00040105
hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate

Specifications

Colorless
3063-94-3
MFCD00040105
hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-hexafluoroisopropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, methacrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, 2,2,2-trifluoro-1-trifluoromethyl ethyl methacrylate, hexafluoro-2-propyl methacrylate, 1h-1-trifluoromethyl trifluoroethyl methacrylate, 1,1,1,3,3,3-hexafluoroprop-2-yl 2-methylprop-2-enoate, 1,1,1,3,3,3-hexafluoro-2-propanyl methacrylate, hfipma
CC(=C)C(=O)OC(C(F)(F)F)C(F)(F)F
236.113
236.11
≥98.0% (GC)
10g
C7H6F6O2
3272
FMQPBWHSNCRVQJ-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoropropan-2-yl 2-methylprop-2-enoate
76469
Clear Liquid at 20°C
1,1,1,3,3,3-Hexafluoroisopropyl Methacrylate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1031

TSCA : Yes