Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-Methoxyphenol

$58.88 - $58.88

Chemical Identifiers

Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl acrylate
CAS 2160-89-6
Name Note stabilized with 50ppm 4-Methoxyphenol
Molecular Formula C6H4F6O2
Linear Formula CH2=CHCO2CH(CF3)2
Formula Weight 222.09g/mol
MDL Number MFCD00040104
UN Number UN3272
Beilstein 2094742
Synonym Acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2139606

 
alfa aesar™
B21396-06
5g Each for $58.88
Description & Specifications

Specifications

Boiling Point 73° to 75°C
Quantity 5g
Flash Point 10°C (50°F)
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl acrylate
Name Note stabilized with 50ppm 4-Methoxyphenol
CAS 2160-89-6
Assay ≥98%
Molecular Formula C6H4F6O2
Linear Formula CH2=CHCO2CH(CF3)2
Formula Weight 222.09g/mol
MDL Number MFCD00040104
UN Number UN3272
Beilstein 2094742
Synonym Acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.