Please login to your online account to display your discounted pricing

1,1,1,3,3,3-Hexafluoroisopropanol-d2, 99+ atom% D, ACROS Organics™

$91.12 - $91.12

Chemical Identifiers

Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropanol-d2
Name Note 99+ atom% D
CAS 38701-74-5
Molecular Formula C3D2F6O
Linear Formula CF3CD(OD)CF3
Formula Weight 170.05
MDL Number MFCD00037559
Synonym 1,1,1,3,3,3-Hexafluoro-2-propanol-d
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC278800010

  SDS
thermo fisher scientific
278800010
1g Each for $91.12
Description & Specifications

Specifications

Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropanol-d2
Name Note 99+ atom% D
CAS 38701-74-5
Deuterium (D) 99.0% min.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 254-091-9
TSCA TSCA
Density 1.6400g/mL
Physical Form Liquid
Quantity 1g
Color Clear colorless
Molecular Formula C3D2F6O
Linear Formula CF3CD(OD)CF3
Formula Weight 170.05
MDL Number MFCD00037559
Synonym 1,1,1,3,3,3-Hexafluoro-2-propanol-d
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.