Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoroacetone trideuterate, 99.5% (Isotopic)

Chemical Identifiers

Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone trideuterate
CAS 109640-39-3
Name Note Isotopic
Formula Weight 226.10 (166.02 Anhydrous)
MDL Number MFCD00149064
UN Number UN2552
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4229009

  SDS
alfa aesar™
4229009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone trideuterate
Name Note Isotopic
CAS 109640-39-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAFLUOROACETONE HYDRATE, LIQUID
Sensitivity Moisture Sensitive
Density 1.71g/mL
Melting Point 21°C
Physical Form Liquid
Assay 99.5%
Quantity 10g
Formula Weight 226.10 (166.02 Anhydrous)
MDL Number MFCD00149064
UN Number UN2552
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.