Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoro-2-propanol-d2, 98%(Isotopic)

Chemical Identifiers

Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol-d2
CAS 38701-74-5
Name Note Isotopic
Formula Weight 170.05
MDL Number MFCD00037559
UN Number UN1760
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4229103

  SDS
alfa aesar™
4229103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4229146

  SDS
alfa aesar™
4229146
5 x 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.05
Packaging Prescored Ampules
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol-d2
Sensitivity Moisture Sensitive
Density 1.60g/mL
Name Note Isotopic
Boiling Point 59°C
Melting Point −40°C
Physical Form Liquid
Assay 98%
Quantity 1g
MDL Number MFCD00037559
CAS 38701-74-5
UN Number UN1760
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.