Learn More
1,1,1,3,3,3-Hexafluoro-2-propanol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1274722
Description
1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.
Solubility
Miscible with water, chloroform, ether and acetone.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases, aluminum, zinc and alkali metals.
Specifications
1,1,1,3,3,3-Hexafluoro-2-propanol | |
-4°C | |
59°C to 60°C | |
1.275 | |
MFCD00011651 | |
UN1760 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol | |
13529 | |
168.04 |
920-66-1 | |
1.619 | |
C3H2F6O | |
CF3CH(OH)CF3 | |
100 g | |
1841007 | |
Miscible with water,chloroform,ether and acetone. | |
C(C(F)(F)F)(C(F)(F)F)O | |
168.038 | |
CHEBI:63104 | |
≥99% |
Chemical Identifiers
920-66-1 | |
168.038 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
13529 | |
1,1,1,3,3,3-hexafluoropropan-2-ol |
C3H2F6O | |
MFCD00011651 | |
1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol | |
CHEBI:63104 | |
C(C(F)(F)F)(C(F)(F)F)O |
Safety and Handling
GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 213-059-4
RTECSNumber : UB6450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only