missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol 99.0+%, TCI America™
Supplier: TCI America H072425G
Specifications
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol | |
Colorless | |
C9H6F6O | |
25 g | |
1,1,1,3,3,3-hexafluoro-2-phenyl-2-propanol, hexafluoro-2-hydroxyisopropyl benzene, bis trifluoromethyl phenylcarbinol, 2,2,2,2',2',2'-hexafluorocumyl alcohol, alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2-phenylisopropanol, benzenemethanol, .alpha.,.alpha.-bis trifluoromethyl, hexafluoro-2-phenyl isopropanol, hexafluoro-2-phenyl-2-propanol, benzenemethanol, alpha,alpha-bis trifluoromethyl | |
C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)O | |
244.136 | |
244.14 | |
Liquid |
718-64-9 | |
160°C | |
MFCD00040983 | |
1987 | |
IZPIZCAYJQCTNG-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol | |
69748 | |
≥99.0% (GC) |
Chemical Identifiers
718-64-9 | |
244.136 | |
IZPIZCAYJQCTNG-UHFFFAOYSA-N | |
69748 | |
C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)O |
C9H6F6O | |
MFCD00040983 | |
1,1,1,3,3,3-hexafluoro-2-phenyl-2-propanol, hexafluoro-2-hydroxyisopropyl benzene, bis trifluoromethyl phenylcarbinol, 2,2,2,2',2',2'-hexafluorocumyl alcohol, alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2-phenylisopropanol, benzenemethanol, .alpha.,.alpha.-bis trifluoromethyl, hexafluoro-2-phenyl isopropanol, hexafluoro-2-phenyl-2-propanol, benzenemethanol, alpha,alpha-bis trifluoromethyl | |
1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol |
Safety and Handling
TSCA : Yes