Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol, 98+%

Chemical Identifiers

Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol
CAS 718-64-9
Molecular Formula C9H6F6O
Formula Weight 244.14
MDL Number MFCD00040983
UN Number UN1987
Beilstein 2053547
Synonym alpha,alpha-Bis(trifluoromethyl)benzyl alcohol; Hexafluorocumyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2136603

  SDS
alfa aesar™
B2136603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2136614

  SDS
alfa aesar™
B2136614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol
Flash Point 60°C (140°F)
Boiling Point 159° to 160°C
Quantity 1g
CAS 718-64-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA TSCA
Assay ≥98%
Molecular Formula C9H6F6O
Formula Weight 244.14
MDL Number MFCD00040983
UN Number UN1987
Beilstein 2053547
Synonym alpha,alpha-Bis(trifluoromethyl)benzyl alcohol; Hexafluorocumyl alcohol
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.