missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,2,2-Pentafluoro-3-iodopropane, 96%

Manufacturer:  Alfa Aesar™ L1104706

 View more versions of this product

Catalog No. AAL1104706



Description

Description

Applications
1,1,1,2,2-Pentafluoro-3-iodopropane is used to produce 1,1,1,2,2-pentafluoro-propane by heating. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,1,1,2,2-Pentafluoro-3-iodopropane
5g
C3H2F5I
1700278
HENALDZJQYAUBN-UHFFFAOYSA-N
1,1,1,2,2-pentafluoro-3-iodopropane
67721
96%
70°C to 71°C
354-69-8
MFCD00039405
pentafluoropropyl iodide, 2,2,3,3,3-pentafluoropropyl iodide, propane, 1,1,1,2,2-pentafluoro-3-iodo, 1h,1h-pentafluoropropyl iodide, 1-iodo-2,2,3,3,3-pentafluoropropane, acmc-1cmxs, cf3cf2ch2i, 2,3,3,3-pentafluoropropyl iodide, 3-iodo-1,1,1,2,2-pentafluoropropane, propane,1,1,2,2-pentafluoro-3-iodo
C(C(C(F)(F)F)(F)F)I
259.945
259.95

Chemical Identifiers

354-69-8
259.945
HENALDZJQYAUBN-UHFFFAOYSA-N
67721
C(C(C(F)(F)F)(F)F)I
C3H2F5I
MFCD00039405
pentafluoropropyl iodide, 2,2,3,3,3-pentafluoropropyl iodide, propane, 1,1,1,2,2-pentafluoro-3-iodo, 1h,1h-pentafluoropropyl iodide, 1-iodo-2,2,3,3,3-pentafluoropropane, acmc-1cmxs, cf3cf2ch2i, 2,3,3,3-pentafluoropropyl iodide, 3-iodo-1,1,1,2,2-pentafluoropropane, propane,1,1,2,2-pentafluoro-3-iodo
1,1,1,2,2-pentafluoro-3-iodopropane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents