Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione, 97%

Chemical Identifiers

Chemical Name or Material 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione
CAS 17587-22-3
Molecular Formula C10H11F7O2
Formula Weight 296.19
MDL Number MFCD00000435
UN Number UN1224
Beilstein 2056991
Synonym 7,7-Dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedione; Fod-H
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1207706

 
alfa aesar™
A1207706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1207714

 
alfa aesar™
A1207714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1207722

 
alfa aesar™
A1207722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 296.19
Chemical Name or Material 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione
TSCA TSCA
Assay 97%
Molecular Formula C10H11F7O2
MDL Number MFCD00000435
CAS 17587-22-3
UN Number UN1224
Beilstein 2056991
Synonym 7,7-Dimethyl-1,1,1,2,2,3,3-heptafluoro-4,6-octanedione; Fod-H
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Recommended Storage 2°C to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Flash Point 53°C (127°F)
Boiling Point 56° to 57°C (10mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2°C to 8°C