Please login to your online account to display your discounted pricing

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC312440250

 View more versions of this product

Catalog No. AC312440250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 474
Physical Form Liquid
Quantity 25g
Color Clear
Density 1.9340g/mL
CAS Min % 96.0
CAS Max % 100.0
Linear Formula ICH2CH2(CF2)5CF3
Synonym 1H,1H,2H,2H-Perfluorooctyl iodide
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 2043-57-4
Chemical Name or Material 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane
Molecular Formula C8H4F13I
MDL Number MFCD00039410
Packaging Glass bottle
Refractive Index 1.3590 to 1.3610 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane
CAS 2043-57-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H4F13I
Linear Formula ICH2CH2(CF2)5CF3
Formula Weight 474
MDL Number MFCD00039410
Synonym 1H,1H,2H,2H-Perfluorooctyl iodide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents