Please login to your online account to display your discounted pricing

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane
CAS 2043-57-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H4F13I
Linear Formula ICH2CH2(CF2)5CF3
Formula Weight 474
MDL Number MFCD00039410
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC312440250

  SDS
thermo fisher scientific
AC312440250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 474
Quantity 25g
Color Clear
Density 1.9340g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane
Assay Percent Range 97%
Molecular Formula C8H4F13I
Linear Formula ICH2CH2(CF2)5CF3
Infrared Spectrum Authentic
MDL Number MFCD00039410
CAS 2043-57-4
Synonym 1H,1H,2H,2H-Perfluorooctyl iodide
Packaging Glass bottle
Refractive Index 1.3590 to 1.3610 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.