Thermo Scientific™ 0.8 mL Storage Plate
0.8 mL deepwell storage plates for cell culture or sample storage applications.
$561.79 - $695.00
Specifications
Certifications/Compliance Certified free from human DNA, RNases, and DNases.
Well Design Conical
No. per Pack 50
Volume (Metric) Well 800µL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
AB0765W Thermo Scientific™
AB0765W
Case of 50 Each for $695.00
AB0765K Thermo Scientific™
AB0765K
Case of 50 Each for $689.06
AB-0765 Thermo Scientific™
AB0765
Case of 50 Each for $561.79
Specifications
Description
Streamline your molecular biology applications with Thermo Scientific Storage and Assay Plates. All plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored or transported easily. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
  • 0.8 mL maximum well volume when sealed with adhesive or heat seals
  • 0.70 mL working volume when sealed with Storage Plate Cap Strips
  • 0.55 mL working volume when sealed with a 96-well Sealing Mat
  • Conical well bottom for ease of sample retrieval
Description & Specifications
Specifications
Certified free from human DNA, RNases, and DNases.
Conical
50
800µL