Please login to your online account to display your discounted pricing

Thermo Scientific™ 0.8 mL Storage Plate


0.8 mL deepwell storage plates for cell culture or sample storage applications.

Manufacturer: thermo scientific™  AB0765W

 View more versions of this product

Catalog No. AB0765W

  • / Case of 50 Each
 

Description & Specifications

Specifications

Certifications/Compliance Certified free from human DNA, RNases, and DNases.
Color White
No. per Pack 50
Volume (Metric) Well 800µL
Well Design Conical

Streamline your molecular biology applications with Thermo Scientific Storage and Assay Plates. All plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored or transported easily. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.

  • 0.8 mL maximum well volume when sealed with adhesive or heat seals
  • 0.70 mL working volume when sealed with Storage Plate Cap Strips
  • 0.55 mL working volume when sealed with a 96-well Sealing Mat
  • Conical well bottom for ease of sample retrieval

ABgene storage and assay plates are manufactured and in a Class 100,000 cleanroom environment to ensure that each plate is contamination-free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low-throughput applications.

  • 96 Round wells with 800μL maximum well volume
  • Max working volume: Film sealing = 0.8mL, Individual Cap Sealing = 0.7mL, Mat Cap Sealing = 0.65mL
  • Raised rim design around each well for optimal sealing and reduced risk of cross-contamination
  • Suitable for use with DMSO, Ethanol, Isopropanol
  • Alphanumeric grid-referencing
  • Designed according to SBS footprint standards for robotic compatibility
  • Wide working temperature range from -196° to +121°C (-321° to +250°F)
  • Can be bar coded for improved sample tracking, please inquire
  • Autoclavable (15 min. at 121°C), safe for gamma irradiation sterilization
  • Max RCF: 200xG (0.65mL/well)
  • Manufactured from virgin medical grade polypropylene and certified RNase, DNase and endotoxin free

Compatible with:

Sealing options include: Adhesive Sealing (0.8mL working volume); Heat sealing (0.8mL working volume); Storage Plate Cap Strips (0.75mL working volume); 96 Cap Sealing Mats (0.65mL working volume)