missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Features chemical resistant O-ring
Manufacturer: Axygen™ SCT050AS
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Available in range of colors for easy sample identification
Specifications
RNase, DNase, pyrogen-free | |
Polypropylene | |
Polypropylene | |
Non-looped | |
Bag | |
No | |
Conical |
Assorted | |
0.5 mL | |
20000xG | |
O-ring | |
Sterile | |
Screw Cap |